accessibility menu, dialog, popup

UserName

Viewing profile as user:
A(a)

CAS RN 32354-50-0

IUPAC Name: —
Synonyms: λ¹-rhodium(1+) norbornadiene (2Z)-4-oxopent-2-en-2-olate
Molecular Weight (g/mol): 294.16
Molecular Formula: C12H15O2Rh
InChi Key: —
SMILES: —
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)