Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Acetylacetonato)(norbornadiene)rhodium(I), TCI America™
SDP

Catalog No. A2761200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2761200MG 200 mg
A27611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A2761200MG Supplier TCI America Supplier No. A2761200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32354-50-0
Molecular Formula C12H15O2Rh
Molecular Weight (g/mol) 294.16
MDL Number MFCD00075071
InChI Key IIQPRGKBFZGPIB-LWFKIUJUSA-M
Synonym (Acetylacetonato)[bicyclo[2.2.1]hepta-2,5-diene]rhodium(I)
IUPAC Name λ¹-rhodium(1+) bicyclo[2.2.1]hepta-2,5-diene (2Z)-4-oxopent-2-en-2-olate
SMILES [Rh+].C\C([O-])=C\C(C)=O.C1C2C=CC1C=C2

Specifications

Melting Point 175°C
Quantity 200 mg
Formula Weight 294.16
Physical Form Crystalline Powder
Chemical Name or Material (Acetylacetonato)(norbornadiene)rhodium(I)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.