accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 32385-06-1

IUPAC Name: 4-amino-6-methoxy-2-methyloxan-3-ol hydrochloride
Synonyms: 4-amino-6-methoxy-2-methyloxan-3-ol hydrochloride
Molecular Weight (g/mol): 197.66
Molecular Formula: C7H16ClNO3
InChi Key: JVYGBUAXDCLPMP-UHFFFAOYNA-N
SMILES: Cl.COC1CC(N)C(O)C(C)O1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results