accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOHNNNOOPOOHHOHOHOBa

CAS RN 3249-92-1

IUPAC Name: {[3,4-dihydroxy-5-(6-oxo-6,9-dihydro-3H-purin-9-yl)oxolan-2-yl]methoxy}phosphonic acid barium
Synonyms: inosinic acid barium
Molecular Weight (g/mol): 485.54
Molecular Formula: C10H13BaN4O8P
InChi Key: JKIAOYFSNWPEEW-UHFFFAOYNA-N
SMILES: [Ba].OC1C(COP(O)(O)=O)OC(C1O)N1C=NC2=C1NC=NC2=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results