accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 325457-99-6

IUPAC Name: 4-chloro-N-(6-chloropyridin-3-yl)benzamide
Synonyms: 4-chloro-N-(6-chloropyridin-3-yl)benzamide
Molecular Weight (g/mol): 267.11
Molecular Formula: C12H8Cl2N2O
InChi Key: FOYOGIUAMYSVIT-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)NC1=CN=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results