accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COClOOH

CAS RN 3260-93-3

IUPAC Name: 3-chloro-2-methoxybenzoic acid
Synonyms: 3-chloro-2-methoxybenzoic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: NKVUYEGKHRDEBB-UHFFFAOYSA-N
SMILES: COC1=C(C=CC=C1Cl)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results