accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3266-23-7

IUPAC Name: 2,3-dimethyloxirane
Synonyms: butylene oxide
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: PQXKWPLDPFFDJP-UHFFFAOYNA-N
SMILES: CC1OC1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results