CAS RN 326914-10-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ SU11652
A cell-permeable pyrrole-indolinone compound that acts as a potent, reversible, and ATP-competitive tyrosine kinase receptor and angiogenic inhibitor.