accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFBrF

CAS RN 327-52-6

IUPAC Name: 1-bromo-2,4,5-trifluorobenzene
Synonyms: 1-bromo-2,4,5-trifluorobenzene
Molecular Weight (g/mol): 210.98
Molecular Formula: C6H2BrF3
InChi Key: DVTULTINXNWGJY-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(Br)C=C1F
Boiling Point 
  • (2)
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)