Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Bromo-2,4,5-trifluorobenzene, 98%, Thermo Scientific™

Catalog No. AC271160250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC271160250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC271160250 Supplier Thermo Scientific Chemicals Supplier No. 271160250

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 327-52-6
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
InChI Key DVTULTINXNWGJY-UHFFFAOYSA-N
Synonym 2,4,5-trifluorobromobenzene, 2-bromo-1,4,5-trifluorobenzene, benzene, 1-bromo-2,4,5-trifluoro, 1-bromo-2,4,5-trifluoro-benzene, paragos 390205, 5-bromo-1,2,4-trifluorobenzene, fr bf df ee, pubchem2306, acmc-1cs6j, ksc224g3h
PubChem CID 67597
IUPAC Name 1-bromo-2,4,5-trifluorobenzene
SMILES C1=C(C(=CC(=C1F)Br)F)F

Specifications

Melting Point -19.0°C
Density 1.8020g/mL
Boiling Point 143.0°C to 144.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Refractive Index 1.4850 to 1.4870
Linear Formula BrC6H2F3
Quantity 25 g
Beilstein 05,679
Specific Gravity 1.802
Solubility Information Solubility in water: insoluble.
Formula Weight 210.98
Percent Purity 98%
Chemical Name or Material 1-Bromo-2,4,5-trifluorobenzene
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and ea
EINECSNumber 206-318-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.