accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NONOS

CAS RN 327036-89-5

IUPAC Name: 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
Synonyms: 4-benzyl-2-methyl-1,2,4-thiadiazolidine-3,5-dione
Molecular Weight (g/mol): 222.26
Molecular Formula: C10H10N2O2S
InChi Key: JDSJDASOXWCHPN-UHFFFAOYSA-N
SMILES: CN1SC(=O)N(CC2=CC=CC=C2)C1=O
Product Type 
  • (3)
Color 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results