accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NN

CAS RN 32741-11-0

IUPAC Name: 2-tert-butylpyrazine
Synonyms: 2-tert-butylpyrazine
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: YNQZVOWNJKASKQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CN=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results