accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNBrBr

CAS RN 32779-37-6

IUPAC Name: 2,5-dibromopyrimidine
Synonyms: 2,5-dibromopyrimidine
Molecular Weight (g/mol): 237.88
Molecular Formula: C4H2Br2N2
InChi Key: XAHITOJPIWZJHD-UHFFFAOYSA-N
SMILES: BrC1=CN=C(Br)N=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results