accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2O

CAS RN 32783-20-3

IUPAC Name: [(prop-1-en-2-yloxy)methyl]benzene
Synonyms: [(prop-1-en-2-yloxy)methyl]benzene
Molecular Weight (g/mol): 148.21
Molecular Formula: C10H12O
InChi Key: ZIFHFFOAEFKJJL-UHFFFAOYSA-N
SMILES: CC(=C)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results