accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 328076-61-5

IUPAC Name: 3-[3-(4-methoxyphenyl)prop-2-enoyl]-6-methyl-4-phenyl-1,2-dihydroquinolin-2-one
Synonyms: 3-[3-(4-methoxyphenyl)prop-2-enoyl]-6-methyl-4-phenyl-1H-quinolin-2-one
Molecular Weight (g/mol): 395.46
Molecular Formula: C26H21NO3
InChi Key: OJMCCNQNCSCAJV-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=CC(=O)C2=C(C3=CC=CC=C3)C3=CC(C)=CC=C3NC2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results