CAS RN 32828-81-2
IUPAC Name:
4-methoxy-N1,N3-bis[(pyridin-3-yl)methyl]benzene-1,3-dicarboxamide
Synonyms:
picotamide
Molecular Weight (g/mol):
376.42
Molecular Formula:
C21H20N4O3
InChi Key:
KYWCWBXGRWWINE-UHFFFAOYSA-N
SMILES:
COC1=CC=C(C=C1C(=O)NCC1=CC=CN=C1)C(=O)NCC1=CC=CN=C1
1
–
1
of
1
results
1
Thermo Scientific™ Picotamide, 98%
CAS: 32828-81-2 Molecular Formula: C21H10N4O3 Synonym: 4-Methoxy-N,N'-bis(3-pyridylmethyl)isophthalamide;