Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Picotamide, 98%

Catalog No. AAJ67232MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67232MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67232MA Supplier Thermo Scientific™ Supplier No. J67232MA

missing translation for 'validMainframeMsg'

CAS: 32828-81-2 | C21H20N4O3, C21H10N4O3 | 376.42 g/mol

 

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32828-81-2
Molecular Formula C21H10N4O3
Synonym 4-Methoxy-N,N'-bis(3-pyridylmethyl)isophthalamide;

Specifications

Quantity 10 mg
Description White
Formula Weight 366.34
Melting Point 128°C to 131°C
Product Type Picotamide, 98%
Form Solid
Assay 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.