accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOO

CAS RN 32953-14-3

IUPAC Name: N-ethyl-2H-1,3-benzodioxol-5-amine
Synonyms: N-ethyl-2H-1,3-benzodioxol-5-amine
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: FPKGTVXPIULTIP-UHFFFAOYSA-N
SMILES: CCNC1=CC=C2OCOC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results