accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOFFFFF

CAS RN 32974-36-0

IUPAC Name: (2,3,4,5,6-pentafluorophenyl)methyl 4-methylbenzene-1-sulfonate
Synonyms: (2,3,4,5,6-pentafluorophenyl)methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 352.28
Molecular Formula: C14H9F5O3S
InChi Key: BKNSDBYJUGNUDL-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC1=C(F)C(F)=C(F)C(F)=C1F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results