Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentafluorobenzyl p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. T12045G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12045G Supplier TCI America Supplier No. T12045G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 32974-36-0
Molecular Formula C14H9F5O3S
Molecular Weight (g/mol) 352.275
MDL Number MFCD06248628
InChI Key BKNSDBYJUGNUDL-UHFFFAOYSA-N
Synonym PFB - Tosylate, p-Toluenesulfonic Acid Pentafluorobenzyl Ester
PubChem CID 2775939
IUPAC Name (2,3,4,5,6-pentafluorophenyl)methyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC2=C(C(=C(C(=C2F)F)F)F)F

Specifications

Melting Point 78°C
Color White
Quantity 5 g
Formula Weight 352.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pentafluorobenzyl p-Toluenesulfonate [Derivatizing Reagent for GC of Inorganic Anions]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.