accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 329907-28-0

IUPAC Name: 3-{[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanyl}propanoic acid
Synonyms: 3-{[5-(4-methylphenyl)thieno[2,3-d]pyrimidin-4-yl]sulfanyl}propanoic acid
Molecular Weight (g/mol): 330.42
Molecular Formula: C16H14N2O2S2
InChi Key: RAOULLCLLOGTDA-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CSC2=NC=NC(SCCC(O)=O)=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results