accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH3COH3C

CAS RN 33077-87-1

IUPAC Name: (2,4-dimethoxyphenyl)(2-methoxyphenyl)methanone
Synonyms: (2,4-dimethoxyphenyl)(2-methoxyphenyl)methanone
Molecular Weight (g/mol): 272.3
Molecular Formula: C16H16O4
InChi Key: FUGHBBQOPVKADC-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C=C1)C(=O)C1=CC=CC=C1OC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results