accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33089-37-1

IUPAC Name: 1,3,6,9,12,15,18,21-octaoxacyclotetracosane
Synonyms: 1,3,6,9,12,15,18,21-octaoxacyclotetracosane
Molecular Weight (g/mol): 352.42
Molecular Formula: C16H32O8
InChi Key: WHLFCESGIBJNGL-UHFFFAOYSA-N
SMILES: C1COCCOCCOCCOCCOCCOCCOCOC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results