Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

24-Crown 8-Ether 95.0+%, TCI America™
SDP

Catalog No. C17361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C17361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C17361G Supplier TCI America Supplier No. C17361G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33089-37-1
Molecular Formula C16H32O8
Molecular Weight (g/mol) 352.424
MDL Number MFCD00054528
InChI Key BGYBONWLWSMGNV-UHFFFAOYSA-N
Synonym 1,4,7,10,13,16,19,22-Octaoxacyclotetracosane
PubChem CID 10893545
IUPAC Name 1,4,7,10,13,16,19,22-octaoxacyclotetracosane
SMILES C1COCCOCCOCCOCCOCCOCCOCCO1

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 352.42
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 24-Crown 8-Ether
RTECSNumber RH1670000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.