accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNBrNHOH

CAS RN 330981-72-1

IUPAC Name: 2-{[2-(4-bromophenyl)-6-methylpyrimidin-4-yl]amino}ethan-1-ol
Synonyms: 2-{[2-(4-bromophenyl)-6-methylpyrimidin-4-yl]amino}ethanol
Molecular Weight (g/mol): 308.18
Molecular Formula: C13H14BrN3O
InChi Key: DUKPGOOUJNUIOI-UHFFFAOYSA-N
SMILES: CC1=CC(NCCO)=NC(=N1)C1=CC=C(Br)C=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results