accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33100-27-5

IUPAC Name: 1,4,7,10,13-pentaoxacyclopentadecane
Synonyms: 15-crown-5 ether
Molecular Weight (g/mol): 220.27
Molecular Formula: C10H20O5
InChi Key: VFTFKUDGYRBSAL-UHFFFAOYSA-N
SMILES: C1COCCOCCOCCOCCO1
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (5)
Quantity 
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)
  • (4)