Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 15-Crown-5

Catalog No. 1888325G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-883-25G 5 g
18-883-225G 25 g
18-883-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 18-883-25G Supplier Sigma Aldrich Supplier No. 1888325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

15-Crown-5 is a crown ether generally used as a ligand in coordination chemistry because of its strong chelating property with certain alkali cations to form complexes. Some derivatives of 15-crown-5 are used as sensors and probes in different physical-chemical processes, phase-transfer reactions, and selective capture of ions for separation and transport.15-Crown-5 is a crown ether that is generally used as a ligand in coordination chemistry. It shows the ability to complexes with alkali metal ions.

Specifications

CAS 33100-27-5
Density 1.113 g/mL at 20°C
Boiling Point 93°C to 96°C (0.05 mmHg, lit.)
Molecular Formula C10H20O5
Linear Formula C10H20O5
Refractive Index n20/D 1.465
MDL Number MFCD00005110
Quantity 5 g
Synonym 15C5; 1,4,7,10,13-Pentaoxacyclopentadecane
Molecular Weight (g/mol) 220.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.