accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3312-60-5

IUPAC Name: N1-cyclohexylpropane-1,3-diamine
Synonyms: N1-cyclohexylpropane-1,3-diamine
Molecular Weight (g/mol): 156.27
Molecular Formula: C9H20N2
InChi Key: ITZPOSYADVYECJ-UHFFFAOYSA-N
SMILES: NCCCNC1CCCCC1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)