CAS RN 331253-86-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PDK1/Akt/Flt Dual Pathway Inhibitor
The PDK1/Akt/Flt Dual Pathway Inhibitor, also referenced under CAS 331253-86-2, controls the biological activity of PDK1/Akt/Flt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.