CAS RN 331467-05-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Indirubin-3'-monoxime-5-sulphonic Acid
A potent, reversible, and selective inhibitor of Cdk1 (IC50 = 5 nM) and Cdk5 (IC50 = 7 nM) and glycogen synthase kinase-3β (GSK-3β; IC50 = 80 nM).