CAS RN 331684-05-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Aromatase Inhibitor I
The Aromatase Inhibitor I, also referenced under CAS 331684-05-0, controls the biological activity of Aromatase.