accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 331753-56-1

IUPAC Name: dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
Synonyms: dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate
Molecular Weight (g/mol): 613.83
Molecular Formula: C8H4K2O12Sb2
InChi Key: GUJUCWZGYWASLH-UHFFFAOYNA-J
SMILES: [K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)