CAS RN 33183-07-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cyclohexenylferrocene 95.0+%, TCI America™
CAS: 33183-07-2 Molecular Formula: C16H18Fe MDL Number: MFCD00191435 Synonym: Cyclohexenylbis(cyclopentadienyl) Iron, Ferrocenecyclohexen-1-yl