accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNONOOCH3CH3

CAS RN 332382-54-4

IUPAC Name: 2-[2-oxo-2-(2,2,4-trimethyl-1,2,3,4-tetrahydroquinolin-1-yl)ethyl]-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-[2-oxo-2-(2,2,4-trimethyl-3,4-dihydroquinolin-1-yl)ethyl]isoindole-1,3-dione
Molecular Weight (g/mol): 362.43
Molecular Formula: C22H22N2O3
InChi Key: KDDHBJICVBONAX-UHFFFAOYNA-N
SMILES: CC1CC(C)(C)N(C(=O)CN2C(=O)C3=CC=CC=C3C2=O)C2=CC=CC=C12
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results