CAS RN 332922-63-1
IUPAC Name:
7-ethoxy-N3-[(furan-2-yl)methyl]acridine-3,9-diamine
Synonyms:
7-ethoxy-N3-(furan-2-ylmethyl)acridine-3,9-diamine
Molecular Weight (g/mol):
333.39
Molecular Formula:
C20H19N3O2
InChi Key:
PGNWQNJXXLPZEL-UHFFFAOYSA-N
SMILES:
CCOC1=CC2=C(N)C3=CC=C(NCC4=CC=CO4)C=C3N=C2C=C1
1
–
1
of
1
results
1
R&D Systems™ STOCK2S 26016, >98%
CAS: 332922-63-1 Molecular Formula: C20H19N3O2