accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOFFF

CAS RN 333-36-8

IUPAC Name: 1,1,1-trifluoro-2-(2,2,2-trifluoroethoxy)ethane
Synonyms: flurothyl hexafluorodiethyl ether flurothyl (substance) fluorothyl flurotyl 氟替尔 flurotylum flurotilo indoklon
Molecular Weight (g/mol): 182.07
Molecular Formula: C4H4F6O
InChi Key: KGPPDNUWZNWPSI-UHFFFAOYSA-N
SMILES: FC(F)(F)COCC(F)(F)F
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)