accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFFHONH4

CAS RN 3336-58-1

IUPAC Name: ammonium trifluoroacetic acid
Synonyms: ammonium trifluoroacetic acid
Molecular Weight (g/mol): 132.06
Molecular Formula: C2H5F3NO2
InChi Key: YCNIBOIOWCTRCL-UHFFFAOYSA-O
SMILES: [NH4+].OC(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (8)
Quantity 
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
  • (1)
Grade 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (2)
  • (8)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)