Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trifluoroacetic acid, ammonium salt, 98%, Thermo Scientific™

Catalog No. AC418860010
Change view
Click to view available options
Quantity:
4 x 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC418860010 4 x 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC418860010 Supplier Thermo Scientific Chemicals Supplier No. 218860010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3336-58-1
Molecular Formula C2H5F3NO2
Molecular Weight (g/mol) 132.06
MDL Number MFCD00012615,MFCD03095537
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonym Ammonium trifluoroacetate
PubChem CID 86601334
IUPAC Name ammonium trifluoroacetic acid
SMILES [NH4+].OC(=O)C(F)(F)F

Specifications

Melting Point 122.0°C to 126.0°C
Quantity 4 x 1 L
Linear Formula CF3CO2NH4
Solubility Information Solubility in water: 100g/L (25°C).
Formula Weight 131.05
Chemical Name or Material Trifluoroacetic acid, ammonium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.