Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LiChropur™ Ammonium Trifluoroacetate, MilliporeSigma™ Supelco™

Catalog No. 111022674 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2674 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2674 Supplier MilliporeSigma Supelco Supplier No. 5686510G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For HPLC

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3336-58-1
Molecular Formula C2H5F3NO2
Molecular Weight (g/mol) 132.06
MDL Number MFCD00012615,MFCD03095537
InChI Key YCNIBOIOWCTRCL-UHFFFAOYSA-O
Synonym Trifluoroacetic acid ammonium salt
IUPAC Name ammonium trifluoroacetic acid
SMILES [NH4+].OC(=O)C(F)(F)F

Specifications

Melting Point 123°C to 125°C (literature)
pH 5 to 6.5
Linear Formula CF3CO2NH4
Quantity 10 g
Formula Weight 131.05
Percent Purity 99% to 101% (NT)
Grade For HPLC
Hazard Category H315 - H319 - H335

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.