CAS RN 3343-39-3
IUPAC Name:
dihydrogen 2-(pyridin-2-yl)ethan-1-amine dichloride
Synonyms:
dihydrogen 2-pyridylethylamine dichloride
Molecular Weight (g/mol):
195.09
Molecular Formula:
C7H12Cl2N2
InChi Key:
GPIFKSYYILNJNR-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].NCCC1=CC=CC=N1
1
–
1
of
1
results
1
R&D Systems™ 2-Pyridylethylamine dihydrochloride
CAS: 3343-39-3 Molecular Formula: C7H12Cl2N2