CAS RN 33462-80-5
IUPAC Name:
(3-chloropropyl)diphenylsulfanium; tetrafluoroboranuide
Synonyms:
(3-chloropropyl)diphenylsulfanium tetrafluoroborate
Molecular Weight (g/mol):
350.61
Molecular Formula:
C15H16BClF4S
InChi Key:
AVYMGNNILZYVOV-UHFFFAOYSA-N
SMILES:
F[B-](F)(F)F.ClCCC[S+](C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
(3-Chloropropyl)diphenylsulfonium Tetrafluoroborate 98.0+%, TCI America™
CAS: 33462-80-5 Molecular Formula: C15H16BClF4S Molecular Weight (g/mol): 350.607 MDL Number: MFCD00074932 InChI Key: AVYMGNNILZYVOV-UHFFFAOYSA-N PubChem CID: 2736650 IUPAC Name: 3-chloropropyl(diphenyl)sulfanium;tetrafluoroborate SMILES: [B-](F)(F)(F)F.C1=CC=C(C=C1)[S+](CCCCl)C2=CC=CC=C2