CAS RN 33471-33-9
IUPAC Name:
2,3,4,5,6-pentahydroxycyclohexan-1-one
Synonyms:
2,3,4,5,6-pentahydroxycyclohexan-1-one
Molecular Weight (g/mol):
178.14
Molecular Formula:
C6H10O6
InChi Key:
VYEGBDHSGHXOGT-UHFFFAOYNA-N
SMILES:
OC1C(O)C(O)C(=O)C(O)C1O
1
–
1
of
1
results
1
1L-epi-2-Inosose 98.0+%, TCI America™
CAS: 33471-33-9 Molecular Formula: C6H10O6 Molecular Weight (g/mol): 178.14 MDL Number: MFCD00038395 InChI Key: VYEGBDHSGHXOGT-SHRIJZPSSA-N Synonym: 2L-2,3,4,6/5-Pentahydroxycyclohexanone PubChem CID: 9877530 IUPAC Name: (2S,3S,5S,6R)-2,3,4,5,6-pentahydroxycyclohexan-1-one SMILES: C1(C(C(C(=O)C(C1O)O)O)O)O