CAS RN 334827-98-4
IUPAC Name:
5-{2-ethoxy-5-[(4-ethylpiperazin-1-yl)sulfonyl]pyridin-3-yl}-3-ethyl-2-(2-methoxyethyl)-2H,4H,7H-pyrazolo[4,3-d]pyrimidin-7-one; benzenesulfonic acid
Synonyms:
5-[2-ethoxy-5-(4-ethylpiperazin-1-ylsulfonyl)pyridin-3-yl]-3-ethyl-2-(2-methoxyethyl)-4H-pyrazolo[4,3-d]pyrimidin-7-one; benzenesulfonic acid
Molecular Weight (g/mol):
677.79
Molecular Formula:
C29H39N7O8S2
InChi Key:
STFRDYSZKVPPQF-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)C1=CC=CC=C1.CCOC1=NC=C(C=C1C1=NC(=O)C2=NN(CCOC)C(CC)=C2N1)S(=O)(=O)N1CCN(CC)CC1
1
–
1
of
1
results
1
R&D Systems™ Gisadenafil besylate
CAS: 334827-98-4 Molecular Formula: C29H39N7O8S2