CAS RN 3349-06-2
IUPAC Name:
nickel(2+) diformate
Synonyms:
nickel(2+) diformate
Molecular Weight (g/mol):
148.73
Molecular Formula:
C2H2NiO4
InChi Key:
HZPNKQREYVVATQ-UHFFFAOYSA-L
SMILES:
[Ni++].[O-]C=O.[O-]C=O
1
–
1
of
1
results
1
Nickel(II) formate, Thermo Scientific™
CAS: 3349-06-2 Molecular Formula: C2H2NiO4 Molecular Weight (g/mol): 148.73 MDL Number: MFCD00050833 InChI Key: HZPNKQREYVVATQ-UHFFFAOYSA-L