accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3352-87-2

IUPAC Name: N,N-diethyldodecanamide
Synonyms: dodecanamide, N,N-diethyl-
Molecular Weight (g/mol): 255.45
Molecular Formula: C16H33NO
InChi Key: CWNSVVHTTQBGQB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)N(CC)CC
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)