Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyldodecanamide 98.0+%, TCI America™
SDP

Catalog No. D1407500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1407500ML 500 mL
D140725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1407500ML Supplier TCI America Supplier No. D1407500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3352-87-2
Molecular Formula C16H33NO
Molecular Weight (g/mol) 255.446
MDL Number MFCD00008962
InChI Key CWNSVVHTTQBGQB-UHFFFAOYSA-N
Synonym dodecanamide, n,n-diethyl, diethyllauramide, n,n-diethyllauramide, n,n-diethyllaurylamide, nopcogen 14-l, unii-0aj0n90byy, 0aj0n90byy, dodecanamide,n-diethyl, n,n-diethyl lauramide, n-lauroyldiethylamine
PubChem CID 18783
IUPAC Name N,N-diethyldodecanamide
SMILES CCCCCCCCCCCC(=O)N(CC)CC

Specifications

Color Yellow
Boiling Point 167°C
Quantity 500 mL
Formula Weight 255.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethyldodecanamide
EINECSNumber (2)-0806
RTECSNumber RB4200000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.