Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethyldodecanamide 98+%, Thermo Scientific™

Catalog No. AC186472500
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC186472500 250 g
AC186470500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC186472500 Supplier Thermo Scientific Chemicals Supplier No. 186472500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3352-87-2
Molecular Formula C16H33NO
Molecular Weight (g/mol) 255.446
InChI Key CWNSVVHTTQBGQB-UHFFFAOYSA-N
Synonym dodecanamide, n,n-diethyl, diethyllauramide, n,n-diethyllauramide, n,n-diethyllaurylamide, nopcogen 14-l, unii-0aj0n90byy, 0aj0n90byy, dodecanamide,n-diethyl, n,n-diethyl lauramide, n-lauroyldiethylamine
PubChem CID 18783
IUPAC Name N,N-diethyldodecanamide
SMILES CCCCCCCCCCCC(=O)N(CC)CC

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.84
Infrared Spectrum Authentic
Refractive Index 1.4535 to 1.4555 (20°C, 589nm)
Linear Formula CH3(CH2)10CON(C2H5)2
Quantity 250 g
Formula Weight 255.44
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material N, N-Diethyldodecanamide, 98+%
Show More Show Less
EINECSNumber 222-118-3
RTECSNumber RB4200000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.