accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33577-16-1

IUPAC Name: methanesulfinyl(methylsulfanyl)methane
Synonyms: methyl methylthiomethyl sulphoxide
Molecular Weight (g/mol): 124.22
Molecular Formula: C3H8OS2
InChi Key: OTKFCIVOVKCFHR-UHFFFAOYNA-N
SMILES: CSCS(C)=O
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)