Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl (Methylsulfinyl)methyl Sulfide 96.0+%, TCI America™
SDP

Catalog No. M08055G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M08055G Supplier TCI America Supplier No. M08055G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33577-16-1
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.22
MDL Number MFCD00002091
InChI Key OTKFCIVOVKCFHR-UHFFFAOYNA-N
Synonym methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
PubChem CID 99129
IUPAC Name methanesulfinyl(methylsulfanyl)methane
SMILES CSCS(C)=O

Specifications

Color Yellow
Boiling Point 93°C
Quantity 5 g
Formula Weight 124.22
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Methyl (Methylsulfinyl)methyl Sulfide
EINECSNumber (2)-2801
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.