Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl methylsulfinylmethyl sulfide, 97%, Thermo Scientific™

Catalog No. AC176150500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176150500 50 g
AC176150100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC176150500 Supplier Thermo Scientific Chemicals Supplier No. 176150500

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33577-16-1
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.22
MDL Number MFCD00002091
InChI Key OTKFCIVOVKCFHR-UHFFFAOYNA-N
Synonym methyl methylsulfinylmethyl sulfide, methyl methylsulfinyl methyl sulfide, methane, methylsulfinyl methylthio, methyl methylthio methyl sulfoxide, famso, methanesulfinyl-methylsulfanyl-methane, formaldehyde dimethyl mercaptal s-oxide, methylsulfinyl methylthio methane, methyl methylthiomethyl sulphoxide, formaldehyde methyl mercaptal s-oxide
PubChem CID 99129
IUPAC Name methanesulfinyl(methylsulfanyl)methane
SMILES CSCS(C)=O

Specifications

Density 1.22g/mL
Assay Percent Range 97%
Linear Formula CH3SOCH2SCH3
Quantity 50 g
Fieser 04,341; 05,456; 06,390; 08,344; 09,314
Formula Weight 124.22
Chemical Name or Material Methyl methylsulfinylmethyl sulfide, 97%
Hazard Category STENCH
Hazard Statement HYGROSCOPIC
EINECSNumber 251-577-2
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.